missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FUCA1 Protein
A cDNA sequence encoding the FUCA1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11909-50ug
Additional Details : Weight : 0.01000kg
Specifications
FUCA1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
VRRAQPPRRY TPDWPSLDSR PLPAWFDEAK FGVFIHWGVF SVPAWGSEWF WWHWQGEGRP QYQRFMRDNY PPGFSYADFG PQFTARFFHP EEWADLFQAA GAKYVVLTTK HHEGFTNWPS PVSWNWNSKD VGPHRDLVGE LGTALRKRNI RYGLYHSLLE WFHPLYLLDK KNGFKTQHFV SAKTMPELYD LVNSYKPDLI WSDGEWECPD TYWNSTNFLS WLYNDSPVKD EVVVNDRWGQ NCSCHHGGYY NCEDKFKPQS LPDHKWEMCT SIDKFSWGYR RDMALSDVTE ESEIISELVQ TVSLGGNYLL NIGPTKDGLI VPIFQERLLA VGKWLSINGE AIYASKPWRV QWEKNTTSVW YTSKGSAVYA IFLHWPENGV LNLESPITTS TTKITMLGIQ GDLKWSTDPD KGLFISLPQL PPSAVPAEFA WTIKLTGVKH HHHHH | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FUCA1 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |