missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FSH Protein
A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.
£99.15 - £430.00
Specifications
Name | FSH Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15928748
|
enQuireBio™
QP11903-100IU |
100 IU |
£99.15
100 units |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938748
|
enQuireBio™
QP11903-5000IU |
5000 IU |
£430.00
5000 units |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948748
|
enQuireBio™
QP11903-500IU |
500 IU |
£256.00
500 units |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FSH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The FSH was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
Human | |
MKTLQFFFLF CCWKAICCNS CELTNITIAI EKEECRFCIS INTTWCAGYC YTRDLVYKDP ARPKIQKTCT FKELVYETVR VPGCAHHADS LYTYPVATQC HCGKCDSDST DCTVRGLGPS YCSFGEMKE |