missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FSH Protein
A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11903-500IU
Additional Details : Weight : 0.01000kg
Specifications
FSH Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MKTLQFFFLF CCWKAICCNS CELTNITIAI EKEECRFCIS INTTWCAGYC YTRDLVYKDP ARPKIQKTCT FKELVYETVR VPGCAHHADS LYTYPVATQC HCGKCDSDST DCTVRGLGPS YCSFGEMKE |
500 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The FSH was lyophilized with no additives. |