missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FKBP2 Protein
A cDNA sequence encoding the FKBP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10608-1mg
Additional Details : Weight : 0.01000kg
Specifications
2286 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 270 nmoles/min/ug, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
Untagged | |
The FKBP2 solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, and 10% glycerol. |
FKBP2 Protein | |
Research Use Only | |
FKBP2 | |
Recombinant Protein | |
E. coli | |
MATGAEGKRK LQIGVKKRVD HCPIKSRKGD VLHMHYTGKL EDGTEFDSSL PQNQPFVFSL GTGQVIKGWD QGLLGMCEGE KRKLVIPSEL GYGERGAPPK IPGGATLVFE VELLKIERRT EL | |
Greater than 90% as determined by SDS-PAGE. |