missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR4 Protein
A cDNA sequence encoding the FGFR4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11877-5ug
Additional Details : Weight : 0.01000kg
Specifications
2264 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
LEASEEVELE PCLAPSLEQQ EQELTVALGQ PVRLCCGRAE RGGHWYKEGS RLAPAGRVRG WRGRLEIASF LPEDAGRYLC LARGSMIVLQ NLTLITGDSL TSSNDDEDPK SHRDPSNRHS YPQQAPYWTH PQRMEKKLHA VPAGNTVKFR CPAAGNPTPT IRWLKDGQAF HGENRIGGIR LRHQHWSLVM ESVVPSDRGT YTCLVENAVG SIRYNYLLDV LERSPHRPIL QAGLPANTTA VVGSDVELLC KVYSDAQPHI QWLKHIVING SSFGADGFPY VQVLKTADIN SSEVEVLYLR NVSAEDAGEY TCLAGNSIGL SYQSAWLTVL PEEDPTWTAA APEARYTDLE HHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
FGFR4 Protein | |
Research Use Only | |
FGFR4 | |
Human | |
His | |
FGFR4 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |