missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR1 Protein
A cDNA sequence encoding the FGFR1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11874-2ug
Additional Details : Weight : 0.01000kg
Specifications
2260 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
RPSPTLPEQA QPWGAPVEVE SFLVHPGDLL QLRCRLRDDV QSINWLRDGV QLAESNRTRI TGEEVEVQDS VPADSGLYAC VTSSPSGSDT TYFSVNVSDA LPSSEDDDDD DDSSSEEKET DNTKPNRMPV APYWTSPEKM EKKLHAVPAA KTVKFKCPSS GTPNPTLRWL KNGKEFKPDH RIGGYKVRYA TWSIIMDSVV PSDKGNYTCI VENEYGSINH TYQLDVVERS PHRPILQAGL PANKTVALGS NVEFMCKVYS DPQPHIQWLK HIEVNGSKIG PDNLPYVQIL KTAGVNTTDK EMEVLHLRNV SFEDAGEYTC LAGNSIGLSH HSAWLTVLEA LEERPAVMTS PLYLELEHHH HHH | |
Greater than 95.0% as determined by SDS-PAGE. |
FGFR1 Protein | |
Research Use Only | |
FGFR1 | |
Human | |
His | |
The FGFR solution (0.5 mg/1ml) contains phosphate buffered Saline (pH 7.4), and 10% glycerol. |