missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGF22 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5374-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15976009

  • £191.00 / 20µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_065688.1
FGF22 Protein
Research Use Only
FGF22
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
27006
20 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MTPSASRGPR . SYPHLEGDV R. WRRLFSST HF. FLRVDPG GRV. QGTRWR HGQD. SILEI RSVHV. GVVV IKAVSS. GFY VAMNRRG. RL YGSRLYTVDC RFRERIEE. N GHNTYASQR.  WRRRGQPMFL . ALDRRGGPR P. GGRTRRYH LS. AHFLPVL VS
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers