missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF22 Protein
Recombinant Protein
Brand: enQuireBio™ QP5374-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_065688.1 | |
FGF22 Protein | |
Research Use Only | |
FGF22 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
27006 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MTPSASRGPR . SYPHLEGDV R. WRRLFSST HF. FLRVDPG GRV. QGTRWR HGQD. SILEI RSVHV. GVVV IKAVSS. GFY VAMNRRG. RL YGSRLYTVDC RFRERIEE. N GHNTYASQR. WRRRGQPMFL . ALDRRGGPR P. GGRTRRYH LS. AHFLPVL VS | |
Greater than 95% as determined by SDS-PAGE. |