missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF14 / SCA27 Protein
A DNA sequence encoding the Homo sapiens (Human) FGF14 / SCA27, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6043-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004106.1 | |
FGF14 / SCA27 Protein | |
Research Use Only | |
FGF14 | |
Recombinant Protein | |
E. coli | |
MVKPVPLFRR TDFKLLLCNH KDLFFLRVSK LLDCFSPKSM WFLWNIFSKG THMLQCLCGK SLKKNKNPTD PQLKGIVTRL YCRQGYYLQM HPDGALDGTK DDSTNSTLFN LIPVGLRVVA IQGVKTGLYI AMNGEGYLYP SELFTPECKF KESVFENYYV IYSSMLYRQQ ESGRAWFLGL NKEGQAMKGN RVKKTKPAAH FLPKPLEVAM YREPSLHDVG ETVPKPGVTP SKSTSASAIM NGGKPVNKSK TT | |
Greater than 80% as determined by SDS-PAGE. |
2259 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |