missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FETUB Protein
A cDNA sequence encoding the FETUB was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | FETUB Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968638
|
enQuireBio™
QP11868-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948638
|
enQuireBio™
QP11868-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958638
|
enQuireBio™
QP11868-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FETUB Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FETUB filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4. |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
HSHVCGAMSP PQLALNPSAL LSRGCNDSDV LAVAGFALRD INKDRKDGYV LRLNRVNDAQ EYRRGGLGSL FYLTLDVLET DCHVLRKKAW QDCGMRIFFE SVYGQCKAIF YMNNPSRVLY LAAYNCTLRP VSKKKIYMTC PDCPSSIPTD SSNHQVLEAA TESLAKYNNE NTSKQYSLFK VTRASSQWVV GPSYFVEYLI KESPCTKSQA SSCSLQSSDS VPVGLCKGSL TRTHWEKFVS VTCDFFESQA PATGSENSAV NQKPTNLPKV EESQQKNTPP TDSPSKAGPR GSVQYLPDLD DKNSQEKGPQ EAFPVHLDLT TNPQGETLDI SFLFLEPMEE KLVVLPFPKE KARTAECPGP AQNASPLVLP PHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |