missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ferritin heavy chain Protein
A DNA sequence encoding the Homo sapiens (Human) Ferritin heavy chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8397-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002023.2 | |
Ferritin heavy chain Protein | |
Research Use Only | |
FTH1 | |
Recombinant Protein | |
E. coli | |
TTASTSQVRQ NYHQDSEAAI NRQINLELYA SYVYLSMSYY FDRDDVALKN FAKYFLHQSH EEREHAEKLM KLQNQRGGRI FLQDIKKPDC DDWESGLNAM ECALHLEKNV NQSLLELHKL ATDKNDPHLC DFIETHYLNE QVKAIKELGD HVTNLRKMGA PESGLAEYLF DKHTLGDSDN ES | |
Greater than 90% as determined by SDS-PAGE. |
2495 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |