missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FEN1 Protein
A cDNA sequence encoding the FEN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11866-5ug
Additional Details : Weight : 0.01000kg
Specifications
2237 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGIQGLAKLI ADVAPSAIRE NDIKSYFGRK VAIDASMSIY QFLIAVRQGG DVLQNEEGET TSHLMGMFYR TIRMMENGIK PVYVFDGKPP QLKSGELAKR SERRAEAEKQ LQQAQAAGAE QEVEKFTKRL VKVTKQHNDE CKHLLSLMGI PYLDAPSEAE ASCAALVKAG KVYAAATEDM DCLTFGSPVL MRHLTASEAK KLPIQEFHLS RILQELGLNQ EQFVDLCILL GSDYCESIRG IGPKRAVDLI QKHKSIEEIV RRLDPNKYPV PENWLHKEAH QLFLEPEVLD PESVELKWSE PNEEELIKFM CGEKQFSEER IRSGVKRLSK SRQGSTQGRL DDFFKVTGSL SSAKRKEPEP KGSTKKKAKT GAAGKFKRGK | |
Greater than 90.0% as determined by SDS-PAGE. |
FEN1 Protein | |
Research Use Only | |
FEN1 | |
Human | |
Untagged | |
The protein contains 20mM Tris-HCl buffer pH 8.0, 1mM DTT, 10% glycerol. |