missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FEN1 Protein

A cDNA sequence encoding the FEN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11866-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968628

  • £155.00 / 20µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

2237
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGIQGLAKLI ADVAPSAIRE NDIKSYFGRK VAIDASMSIY QFLIAVRQGG DVLQNEEGET TSHLMGMFYR TIRMMENGIK PVYVFDGKPP QLKSGELAKR SERRAEAEKQ LQQAQAAGAE QEVEKFTKRL VKVTKQHNDE CKHLLSLMGI PYLDAPSEAE ASCAALVKAG KVYAAATEDM DCLTFGSPVL MRHLTASEAK KLPIQEFHLS RILQELGLNQ EQFVDLCILL GSDYCESIRG IGPKRAVDLI QKHKSIEEIV RRLDPNKYPV PENWLHKEAH QLFLEPEVLD PESVELKWSE PNEEELIKFM CGEKQFSEER IRSGVKRLSK SRQGSTQGRL DDFFKVTGSL SSAKRKEPEP KGSTKKKAKT GAAGKFKRGK
Greater than 90.0% as determined by SDS-PAGE.
FEN1 Protein
Research Use Only
FEN1
Human
Untagged
The protein contains 20mM Tris-HCl buffer pH 8.0, 1mM DTT, 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers