missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FCGR3A Protein
A cDNA sequence encoding the FCGR3A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11860-HIS-20ug
Additional Details : Weight : 0.01000kg
Specifications
2214 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMRT EDLPKAVVFL EPQWYRVLEK DSVTLKCQGA YSPEDNSTQW FHNESLISSQ ASSYFIDAAT VDDSGEYRCQ TNLSTLSDPV QLEVHIGWLL LQAPRWVFKE EDPIHLRCHS WKNTALHKVT YLQNGKGRKY FHHNSDFYIP KATLKDSGSY FCRGLFGSKN VSSETVNITI TQGLAVSTIS SFFPPGYQ | |
Greater than 90% as determined by SDS-PAGE. |
FCGR3A Protein | |
Research Use Only | |
FCGR3A | |
Human | |
His | |
FCGR3A protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 10% glycerol. |