missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM3C Protein
A cDNA sequence encoding the FAM3C was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11846-1mg
Additional Details : Weight : 0.01000kg
Specifications
FAM3C Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQVFEIKMDA SLGNLFARSA LDTAARSTKP PRYKCGISKA CPEKHFAFKM ASGAANVVGP KICLEDNVLM SGVKNNVGRG INVALANGKT GEVLDTKYFD MWGGDVAPFI EFLKAIQDGT IVLMGTYDDG ATKLNDEARR LIADLGSTSI TNLGFRDNWV FCGGKGIKTK SPFEQHIKNN KDTNKYEGWP EVVEMEGCIP QKQD | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FAM3C solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 200mM NaCl and 20% glycerol. |