missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FADD Protein

A cDNA sequence encoding the FADD was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11839-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938548

  • £87.35 / 2µg

Estimated Shipment
30-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FADD Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The FADD protein solution contains 20mM Tris-HCl, pH 8.0, and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHHGMASMTGGQQ MGRDLYDDDD KDRWGSMDPF LVLLHSVSSS LSSSELTELK FLCLGRVGKR KLERVQSGLD LFSMLLEQND LEPGHTELLR ELLASLRRHD LLRRVDDFEA GAAAGAAPGE EDLCAAFNVI CDNVGKDWRR LARQLKVSDT KIDSIEDRYP RNLTERVRES LRIWKNTEKE NATVAHLVGA LRSCQMNLVA DLVQEVQQAR DLQNRSGAMS PMSWNSDAST SEAS
Videos
SDS
Documents