missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP5 / E-FABP Protein
A DNA sequence encoding the Homo sapiens (Human) FABP5 / E-FABP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6014-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001435.1 | |
FABP5 / E-FABP Protein | |
Research Use Only | |
FABP5 | |
Recombinant Protein | |
E. coli | |
MATVQQLEGR WRLVDSKGFD EYMKELGVGI ALRKMGAMAK PDCIITCDGK NLTIKTESTL KTTQFSCTLG EKFEETTADG RKTQTVCNFT DGALVQHQEW DGKESTITRK LKDGKLVVEC VMNNVTCTRI YEKVE | |
Greater than 80% as determined by SDS-PAGE. |
2171 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |