missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FABP4 Protein

A cDNA sequence encoding the FABP4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11836-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968528

  • £1320.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

2167
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human
CDAFVGTWKL VSSENFDDYM KEVGVGFATR KVAGMAKPNM IISVNGDVIT IKSESTFKNT EISFILGQEF DEVTADDRKV KSTITLDGGV LVHVQKWDGK STTIKRKRED DKLVVECVMK GVTSTRVYER A
Greater than 85.0% as determined by SDS-PAGE.
FABP4 Protein
Research Use Only
FABP4
Human
Untagged
FABP4 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 6.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers