missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP3 Protein
A cDNA sequence encoding the FABP3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11835-5ug
Additional Details : Weight : 0.01000kg
Specifications
FABP3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MVDAFLGTWK LVDSKNFDDY MKSLGVGFAT RQVASMTKPT TIIEKNGDIL TLKTHSTFKN TEISFKLGVE FDETTADDRK VKSIVTLDGG KLVHLQKWDG QETTLVRELI DGKLILTLTH GTAVCTRTYE KEA | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
50mM phosphate borate buffer pH 8. |