missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP12 Protein
A cDNA sequence encoding the FABP12 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11833-10ug
Additional Details : Weight : 0.01000kg
Specifications
FABP12 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMIDQLQ GTWKSISCEN SEDYMKELGI GRASRKLGRL AKPTVTISTD GDVITIKTKS IFKNNEISFK LGEEFEEITP GGHKTKSKVT LDKESLIQVQ DWDGKETTIT RKLVDGKMVV ESTVNSVICT RTYEKVSSNS VSNS | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FABP12 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 0.15M NaCl and 10% glycerol. |