missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP12 Protein
A cDNA sequence encoding the FABP12 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | FABP12 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15988518
|
enQuireBio™
QP11833-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968518
|
enQuireBio™
QP11833-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978518
|
enQuireBio™
QP11833-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FABP12 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FABP12 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 0.15M NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMIDQLQ GTWKSISCEN SEDYMKELGI GRASRKLGRL AKPTVTISTD GDVITIKTKS IFKNNEISFK LGEEFEEITP GGHKTKSKVT LDKESLIQVQ DWDGKETTIT RKLVDGKMVV ESTVNSVICT RTYEKVSSNS VSNS | |
Greater than 90.0% as determined by SDS-PAGE. |