missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP1 Protein
A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11832-1mg
Additional Details : Weight : 0.01000kg
Specifications
FABP1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSFSGKYQLQ SQENFEAFMK AIGLPEELIQ KGKDIKGVSEIVQNGKHFKF TITAGSKVIQ NEFTVGEECE LETMTGEKVK TVVQLEGDNK LVTTFKNIKSVTELNGDIIT NTMTLGDIVF KRISKRI |