missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F9 Protein
A cDNA sequence encoding the F9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10595-500IU
Additional Details : Weight : 0.01000kg
Specifications
F9 Protein | |
Research Use Only | |
The activity per mg was tested and found to be 306.5 PEU/mg. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |
500 IU | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human | |
The Factor-IX was lyophilized from a sterile solution containing 20mM Tris-HCl pH 7.4, 0.1M NaCl and 1mM Benzamidine. |