missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F-box only protein 1

A DNA sequence encoding the Homo sapiens (Human) F-box only protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9935-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049031

  • £1773.00 / 1mg

Estimated Shipment: 19-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036307.2
F-box only protein 1
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
26273
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FBXO3
Recombinant Protein
Yeast
MAAMETETAP LTLESLPTDP LLLILSFLDY RDLINCCYVS RRLSQLSSHD PLWRRHCKKY WLISEEEKTQ KNQCWKSLFI DTYSDVGRYI DHYAAIKKAW DDLKKYLEPR CPRMVLSLKE GAREEDLDAV EAQIGCKLPD DYRCSYRIHN GQKLVVPGLL GSMALSNHYR SEDLLDVDTA AGGFQQRQGL KYCLPLTFCI HTGLSQYIAV EAAEGRNKNE VFYQCPDQMA RNPAAIDMFI IGATFTDWFT SYVKNVVSGG FPIIRDQIFR YVHDPECVAT TGDITVSVST SFLPELSSVH PPHYFFTYRI RIEMSKDALP EKACQLDSRY WRITNAKGDV EEVQGPGVVG EFPIISPGRV YEYTSCTTFS TTSGYMEGYY TFHFLYFKDK IFNVAIPRFH MACPTFRVSI ARLEMGPDEY EEMEEEEEEE EEEDEDDDSA DMDESDEDDE EERRRRVFDV PIRRRRCSRL F
Videos
SDS
Documents
Certificates