missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-box / LRR-repeat protein 18 Protein
A DNA sequence encoding the Homo sapiens (Human) F-box/LRR-repeat protein 18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6024-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
F-box / LRR-repeat protein 18 Protein | |
Research Use Only | |
FBXL18 | |
Recombinant Protein | |
E. coli | |
MASSGEDISN DDDDMHPAAA GMADGVHLLG FSDEILLHIL SHVPSTDLIL NVRRTCRKLA ALCLDKSLIH TVLLQKDYQA SEDKVRQLVK EIGREIQQLS MAGCYWLPGS TVEHVARCRS LVKVNLSGCH LTSLRLSKML SALQHLRSLA IDVSPGFDAS QLSSECKATL SRVRELKQTL FTPSYGVVPC CTSLEKLLLY FEILDRTREG AILSGQLMVG QSNVPHYQNL RVFYARLAPG YINQEVVRLY LAVLSDRTPQ NLHAFLISVP GSFAESGATK NLLDSMARNV VLDALQLPKS WLNGSSLLQH MKFNNPFYFS FSRCTLSGGH LIQQVINGGK DLRSLASLNL SGCVHCLSPD SLLCR | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |