missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC7 Protein
A cDNA sequence encoding the EXOSC7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11822-20ug
Additional Details : Weight : 0.01000kg
Specifications
EXOSC7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASVTLS EAEKVYIVHG VQEDLRVDGR GCEDYRCVEV ETDVVSNTSG SARVKLGHTD ILVGVKAEMG TPKLEKPNEG YLEFFVDCSA SATPEFEGRG GDDLGTEIAN TLYRIFNNKS SVDLKTLCIS PREHCWVLYV DVLLLECGGN LFDAISIAVK AALFNTRIPR VRVLEDEEGS KDIELSDDPY DCIRLSVENV PCIVTLCKIG YRHVVDATLQ EEACSLASLL VSVTSKGVVT CMRKVGKGSL DPESIFEMME TGKRVGKVLH ASLQSVVHKE ESLGPKRQKV GFLG | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
EXOSC7 protein solution (0.5 mg/ml) containing 20mM Phosphate buffer saline (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol. |