missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ESM1 Protein
A cDNA sequence encoding the ESM1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11810-HIS-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
ESM1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSWSNNYAV DCPQHCDSSE CKSSPRCKRT VLDDCGCCRV CAAGRGETCY RTVSGMDGMK CGPGLRCQPS NGEDPFGEEF GICKDCPYGT FGMDCRETCN CQSGICDRGT GKCLKFPFFQ YSVTKSSNRF VSLTEHDMAS GDGNIVREEV VKENAAGSPV MRKWLNPR | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ESM1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea and 10% glycerol. |