missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Epiregulin / EREG Protein
A DNA sequence encoding the Homo sapiens (Human) Epiregulin / EREG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5995-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001423.1 | |
Epiregulin / EREG Protein | |
Research Use Only | |
EREG | |
Recombinant Protein | |
E. coli | |
VAQVSITKCS SDMNGYCLHG QCIYLVDMSQ NYCRCEVGYT GVRCEHFFLT VHQPLSKEYV | |
Greater than 90% as determined by SDS-PAGE. |
2069 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |