missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ephrin A4 Protein

A cDNA sequence encoding the Ephrin A4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11800-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958418

  • £540.60 / 10µg
Estimated Shipment: 07-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1945
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MRLLPLLRTV LWAAFLGSPL RGGSSLRHVV YWNSSNPRLL RGDAVVELGL NDYLDIVCPH YEGPGPPEGP ETFALYMVDW PGYESCQAEG PRAYKRWVCS LPFGHVQFSE KIQRFTPFSL GFEFLPGETY YYISVPTPES SGQCLRLQVS VCCKERKSES AHPVGSPGES GTSGWRGGDT PSPLCLLLLL LLLILRLLRI L
Ephrin A4 Protein
Research Use Only
EFNA4
Human
GST
Ephrin A4 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers