missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ENPP1 Protein

A cDNA sequence encoding the ENPP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11790-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15948378

  • £5730.00 / 1mg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ENPP1 Protein
Research Use Only
Recombinant Protein
Human (HEK293)
ASKPSCAKEV KSCKGRCFER TFGNCRCDAA CVELGNCCLD YQETCIEPEH IWTCNKFRCG EKRLTRSLCA CSDDCKDKGD CCINYSSVCQ GEKSWVEEPC ESINEPQCPA GFETPPTLLF SLDGFRAEYL HTWGGLLPVI SKLKKCGTYT KNMRPVYPTK TFPNHYSIVT GLYPESHGII DNKMYDPKMN ASFSLKSKEK FNPEWYKGEP IWVTAKYQGL KSGTFFWPGS DVEINGIFPD IYKMYNGSVP FEERILAVLQ WLQLPKDERP HFYTLYLEEP DSSGHSYGPV SSEVIKALQR VDGMVGMLMD GLKELNLHRC LNLILISDHG MEQGSCKKYI YLNKYLGDVK NIKVIYGPAA RLRPSDVPDK YYSFNYEGIA RNLSCREPNQ HFKPYLKHFL PKRLHFAKSD RIEPLTFYLD PQWQLALNPS ERKYCGSGFH GSDNVFSNMQ ALFVGYGPGF KHGIEADTFE NIEVYNLMCD LLNLTPAPNN GTHGSLNHLL KNPVYTPKHP KEVHPLVQCP FTRNPRDNLG CSCNPSILPI EDFQTQFNLT VAEEKIIKHE TLPYGRPRVL QKENTICLLS QHQFMSGYSQ DILMPLWTSY TVDRNDSFST EDFSNCLYQD FRIPLSPVHK CSFYKNNTKV SYGFLSPPQL NKNSSGIYSE ALLTTNIVPM YQSFQVIWRY FHDTLLRKYA EERNGVNVVS GPVFDFDYDG RCDSLENLRQ KRRVIRNQEI LIPTHFFIVL TSCKDTSQTP LHCENLDTLA FILPHRTDNS ESCVHGKHDS SWVEELLMLH RARITDVEHI TGLSFYQQRK EPVSDILKLK THLPTFSQED GPKLHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers