missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EMCN Protein

A cDNA sequence encoding the EMCN was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11782-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938348

  • £1165.00 / 100µg

Estimated Shipment
26-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

51705
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EMCN
Human
His
EMCN protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 100mM NaCl.
EMCN Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMNSTGV LEAANNSLVV TTTKPSITTP NTESLQKNVV TPTTGTTPKG TITNELLKMS LMSTATFLTS KDEGLKATTT DVRKNDSIIS NVTVTSVTLP NAVSTLQSSK PKTETQSSIK TTEIPGSVLQ PDASPSKTGT LTSIPVTIPE NTSQSQVIGT EGGKNASTSA TSRSYSS
Videos
SDS
Documents