missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Elongation factor 1-delta Protein

A DNA sequence encoding the Homo sapiens (Human) Elongation factor 1-delta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5960-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968949

  • £1735.33 / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001123525.2
Elongation factor 1-delta Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
1936
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EF1D
Recombinant Protein
E. coli
ATNFLAHEKI WFDKFKYDDA ERRFYEQMNG PVAGASRQEN GASVILRDIA RARENIQKSL AGSSGPGASS GTSGDHGELV VRIASLEVEN QSLRGVVQEL QQAISKLEAR LNVLEKSSPG HRATAPQTQH VSPMRQVEPP AKKPATPAED DEDDDIDLFG SDNEEEDKEA AQLREERLRQ YAEKKAKKPA LVAKSSILLD VKPWDDETDM AQLEACVRSI QLDGLVWGAS KLVPVGYGIR KLQIQCVVED DKVGTDLLEE EITKFEEHVQ SVDIAAFNKI
Videos
SDS
Documents
Certificates
Special Offers

Special Offers