missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3G Protein
A DNA sequence encoding the Homo sapiens (Human) EIF3G, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5972-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_003746.2 | |
EIF3G Protein | |
Research Use Only | |
EIF3G | |
Recombinant Protein | |
E. coli | |
PTGDFDSKPS WADQVEEEGE DDKCVTSELL KGIPLATGDT SPEPELLPGA PLPPPKEVIN GNIKTVTEYK IDEDGKKFKI VRTFRIETRK ASKAVARRKN WKKFGNSEFD PPGPNVATTT VSDDVSMTFI TSKEDLNCQE EEDPMNKLKG QKIVSCRICK GDHWTTRCPY KDTLGPMQKE LAEQLGLSTG EKEKLPGELE PVQATQNKTG KYVPPSLRDG ASRRGESMQP NRRADDNATI RVTNLSEDTR ETDLQELFRP FGSISRIYLA KDKTTGQSKG FAFISFHRRE DAARAIAGVS GFGYDHLILN VEWAKPSTN | |
Greater than 90% as determined by SDS-PAGE. |
8666 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |