missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EGF Protein

A cDNA sequence encoding the EGF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10582-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964318

  • £155.00 / 500µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EGF Protein
Research Use Only
Recombinant Protein
P. pastoris
NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
500 μg
< 1.0 EU per ug protein as determined by the LAL method.
Human
Untagged
Each lyophilized mg contains 0.15M NaCl, 0.025M sodium bicarbonate, pH 7.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers