missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RBX1 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase RBX1, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_055063.1 |
---|---|
Gene ID (Entrez) | 9978 |
Name | E3 ubiquitin-protein ligase RBX1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064370
|
enQuireBio™
QP6590-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084370
|
enQuireBio™
QP6590-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074370
|
enQuireBio™
QP6590-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094370
|
enQuireBio™
QP6590-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004380
|
enQuireBio™
QP6590-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014380
|
enQuireBio™
QP6590-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055063.1 | |
E3 ubiquitin-protein ligase RBX1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9978 | |
Research Use Only | |
RBX1 | |
Recombinant Protein | |
E. coli | |
MAAAMDVDTP SGTNSGAGKK RFEVKKWNAV ALWAWDIVVD NCAICRNHIM DLCIECQANQ ASATSEECTV AWGVCNHAFH FHCISRWLKT RQVCPLDNRE WEFQKYGH | |
Greater than 80% as determined by SDS-PAGE. |