missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RAD18 Protein
A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase RAD18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8224-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_064550.3 | |
E3 ubiquitin-protein ligase RAD18 Protein | |
Research Use Only | |
RAD18 | |
Recombinant Protein | |
E. coli | |
MDSLAESRWP PGLAVMKTID DLLRCGICFE YFNIAMIIPQ CSHNYCSLCI RKFLSYKTQC PTCCVTVTEP DLKNNRILDE LVKSLNFARN HLLQFALESP AKSPASSSSK NLAVKVYTPV ASRQSLKQGS RLMDNFLIRE MSGSTSELLI KENKSKFSPQ KEASPAAKTK ETRSVEEIAP DPSEAKRPEP PSTSTLKQVT KVDCPVCGVN IPESHINKHL DSCLSREEKK ESLRSSVHKR KPLPKTVYNL LSDRDLKKKL KEHGLSIQGN KQQLIKRHQE FVHMYNAQCD ALHPKSAAEI VREIENIEKT RMRLEASKLN ESVMVFTKDQ TEKEIDEIHS KYRKKHKSEF QLLVDQARKG YKKIAGMSQK TVTITKEDES TEKLSSVCMG QEDNMTSVTN HFSQSKLDSP EELEPDREED SSSCIDIQEV LSSSESDSCN SSSSDIIRDL LEEEEAWEAS HKNDLQDTEI SPRQNRRTRA AESAEIEPRN KRNRN | |
Greater than 90% as determined by SDS-PAGE. |
56852 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |