missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase NRDP1 Protein

A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase NRDP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8174-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003071

  • £321.05 / 50µg

Estimated Shipment
07-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001229755.1
E3 ubiquitin-protein ligase NRDP1 Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
10193
Greater than 80% as determined by SDS-PAGE.
Research Use Only
RNF41
Recombinant Protein
E. coli
MGYDVTRFQG DVDEDLICPI CSGVLEEPVQ APHCEHAFCN ACITQWFSQQ QTCPVDRSVV TVAHLRPVPR IMRNMLSKLQ IACDNAVFGC SAVVRLDNLM SHLSDCEHNP KRPVTCEQGC GLEMPKDELP NHNCIKHLRS VVQQQQTRIA ELEKTSAEHK HQLAEQKRDI QLLKAYMRAI RSVNPNLQNL EETIEYNEIL EWVNSLQPAR VTRWGGMIST PDAVLQAVIK RSLVESGCPA SIVNELIENA HERSWPQGLA TLETRQMNRR YYENYVAKRI PGKQAVVVMA CENQHMGDDM VQEPGLVMIF AHGVEEI
Videos
Documents