missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DUSP18 Protein

A cDNA sequence encoding the DUSP18 was constructed and used to recombinantly synthesize the protein.

£99.15 - £155.00

Specifications

Name DUSP18 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15958118
View Documents Promotion Details
enQuireBio™
QP11712-5UG
5 μg
£99.15
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15948118
View Documents Promotion Details
enQuireBio™
QP11712-20UG
20 μg
£155.00
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

DUSP18 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DUSP18 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 40% glycerol, 0.1mM PMSF and 1mM EDTA.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMTAPSC AFPVQFRQPS VSGLSQITKS LYISNGVAAN NKLMLSSNQI TMVINVSVEV VNTLYEDIQY MQVPVADSPN SRLCDFFDPI ADHIHSVEMK QGRTLLHCAA GVSRSAALCL AYLMKYHAMS LLDAHTWTKS CRPIIRPNSG FWEQLIHYEF QLFGKNTVHM VSSPVGMIPD IYEKEVRLMI PL
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications