missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP13 Protein
A cDNA sequence encoding the DUSP13 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | DUSP13 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15938118
|
enQuireBio™
QP11711-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918118
|
enQuireBio™
QP11711-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928118
|
enQuireBio™
QP11711-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DUSP13 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DUSP13 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 2mM DTT and 50mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMDSLQK QDLRRPKIHG AVQASPYQPP TLASLQRLLW VRQAATLNHI DEVWPSLFLG DAYAARDKSK LIQLGITHVV NAAAGKFQVD TGAKFYRGMS LEYYGIEADD NPFFDLSVYF LPVARYIRAA LSVPQGRVLV HCAMGVSRSA TLVLAFLMIC ENMTLVEAIQ TVQAHRNICP NSGFLRQLQV LDNRLGRETG RF | |
Greater than 95% as determined by SDS-PAGE. |