missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DUSP13, isoform A Protein

A DNA sequence encoding the Homo sapiens (Human) DUSP13, isoform A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7728-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050461

  • £335.00 / 100µg

Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001007272.1
DUSP13, isoform A Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
51207
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DUSP13
Recombinant Protein
E. coli
MAETSLPELG GEDKATPCPS ILELEELLRA GKSSCSRVDE VWPNLFIGDA ATANNRFELW KLGITHVLNA AHKGLYCQGG PDFYGSSVSY LGVPAHDLPD FDISAYFSSA ADFIHRALNT PGAKVLVHCV VGVSRSATLV LAYLMLHQRL SLRQAVITVR QHRWVFPNRG FLHQLCRLDQ QLRGAGQS
Videos
SDS
Documents
Certificates