missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DPPA4 Protein
A cDNA sequence encoding the DPPA4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | DPPA4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978078
|
enQuireBio™
QP11699-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968078
|
enQuireBio™
QP11699-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958078
|
enQuireBio™
QP11699-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DPPA4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DPPA4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLRGSAS STSMEKAKGK EWTSTEKSRE EDQQASNQPN SIALPGTSAK RTKEKMSIKG SKVLCPKKKA EHTDNPRPQK KIPIPPLPSK LPPVNLIHRD ILRAWCQQLK LSSKGQKLDA YKRLCAFAYP NQKDFPSTAK EAKIRKSLQK KLKVEKGETS LQSSETHPPE VALPPVGEPP ALENSTALLE GVNTVVVTTS APEALLASWA RISARARTPE AVESPQEASG VRWCVVHGKS LPADTDGWVH LQFHAGQAWV PEKQEGRVSA LFLLPASNFP PPHLEDNMLC PKCVHRNKVL IKSLQWE | |
Greater than 85% as determined by SDS-PAGE. |