missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA polymerase processivity factor Protein

A DNA sequence encoding the Homo sapiens (Human) DNA polymerase processivity factor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9615-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048511

  • £670.00 / 100µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DNA polymerase processivity factor Protein
Research Use Only
U27
Recombinant Protein
Yeast
FFFFKAHKAR VGARTSFLTE MERGSRDHHR DHRDHREHRE TREPPTLAFH MKSWKTINKS LKAFAKLLKE NATVTFTPQP SIIIQSAKNH LVQKLTIQAE CLFLSDTDRF LTKTINNHIP LFESFMNIIS NPEVTKMYIQ HDSDLYTRVL VTASDTCTQA SVPCVHGQEV VRDTGRSPLR IDLDHSTVSD VLKWLSPVTK TKRSGKSDAL MAHIIVQVNP PTIKFVTEMN ELEFSNSNKV IFYDVKNMRF NLSAKNLQQA LSMCAVIKTS CSLRTVAAKD CKLILTSKST LLTVEAFLTQ EQLKEESRFE RMGKQDDGKG DRSHKNEDGS ALASKQEMQY EITNYMVPAK NGTAGSSLFN EKEDSESDDS MHFDYSSNPN PKRQRCVV
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers