missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dihydropyrimidinase-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Dihydropyrimidinase-related protein 2, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_001184222.1 |
---|---|
Gene ID (Entrez) | 1808 |
Name | Dihydropyrimidinase-related protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048651
|
enQuireBio™
QP9750-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088651
|
enQuireBio™
QP9750-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058651
|
enQuireBio™
QP9750-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078651
|
enQuireBio™
QP9750-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098651
|
enQuireBio™
QP9750-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068651
|
enQuireBio™
QP9750-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifica
NP_001184222.1 | |
Dihydropyrimidinase-related protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1808 | |
Research Use Only | |
DPYSL2 | |
Recombinant Protein | |
Yeast | |
MSYQGKKNIP RITSDRLLIK GGKIVNDDQS FYADIYMEDG LIKQIGENLI VPGGVKTIEA HSRMVIPGGI DVHTRFQMPD QGMTSADDFF QGTKAALAGG TTMIIDHVVP EPGTSLLAAF DQWREWADSK SCCDYSLHVD ISEWHKGIQE EMEALVKDHG VNSFLVYMAF KDRFQLTDCQ IYEVLSVIRD IGAIAQVHAE NGDIIAEEQQ RILDLGITGP EGHVLSRPEE VEAEAVNRAI TIANQTNCPL YITKVMSKSS AEVIAQARKK GTVVYGEPIT ASLGTDGSHY WSKNWAKAAA FVTSPPLSPD PTTPDFLNSL LSCGDLQVTG SAHCTFNTAQ KAVGKDNFTL IPEGTNGTEE RMSVIWDKAV VTGKMDENQF VAVTSTNAAK VFNLYPRKGR IAVGSDADLV IWDPDSVKTI SAKTHNSSLE YNIFEGMECR GSPLVVISQG KIVLEDGTLH VTEGSGRYIP RKPFPDFVYK RIKARSRLAE LRGVPRGLYD GPVCEVSVTP KTVTPASSAK TSPAKQQAPP VRNLHQSGFS LSGAQIDDNI PRRTTQRIVA PPGGRANITS LG | |
Greater than 90% as determined by SDS-PAGE. |