missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DESI1 Protein
A cDNA sequence encoding the DESI1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11660-10ug
Additional Details : Weight : 0.01000kg
Specifications
27351 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
DESI1 | |
Human | |
His | |
The DESI1 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |
DESI1 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPPNLY PVKLYVYDLS KGLARRLSPI MLGKQLEGIW HTSIVVHKDE FFFGSGGISS CPPGGTLLGP PDSVVDVGST EVTEEIFLEY LSSLGESLFR GEAYNLFEHN CNTFSNEVAQ FLTGRKIPSY ITDLPSEVLS TPFGQALRPL LDSIQIQPPG GSSVGRPNGQ S |