missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Deoxyguanosine kinase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Deoxyguanosine kinase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7588-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001305788.1 | |
Deoxyguanosine kinase, mitochondrial Protein | |
Research Use Only | |
DGUOK | |
Recombinant Protein | |
E. coli | |
RLSIEGNIAV GKSTFVKLLT KTYPEWHVAT EPVATWQNIQ AAGTQKACTA QSLGNLLDMM YREPARWSYT FQTFSFLSRL KVQLEPFPEK LLQARKPVQI FERSVYSDRY IFAKNLFENG SLSDIEWHIY QDWHSFLLWE FASRITLHGF IYLQASPQVC LKRLYQRARE EEKGIELAYL EQLHGQHEAW LIHKTTKLHF EALMNIPVLV LDVNDDFSEE VTKQEDLMRE VNTFVKNL | |
Greater than 90% as determined by SDS-PAGE. |
1716 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |