missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DDOST / OST48 Protein

A DNA sequence encoding the Homo sapiens (Human) DDOST / OST48, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8493-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004931

  • £1688.38 / 1mg

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_002941.1
DDOST / OST48 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
6184
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DDOST
Recombinant Protein
E. coli
SHLAKVTAEV VLAHLGGGST SRATSFLLAL EPELEARLAH LGVQVKGEDE EENNLEVRET KIKGKSGRFF TVKLPVALDP GAKISVIVET VYTHVLHPYP TQITQSEKQF VVFEGNHYFY SPYPTKTQTM RVKLASRNVE SYTKLGNPTR SEDLLDYGPF RDVPAYSQDT FKVHYENNSP FLTITSMTRV IEVSHWGNIA VEENVDLKHT GAVLKGPFSR YDYQRQPDSG ISSIRSFKTI LPAAAQDVYY RDEIGNVSTS HLLILDDSVE MEIRPRFPLF GGWKTHYIVG YNLPSYEYLY NLGDQYALKM RFVDHVFDEQ VIDSLTVKII LPEGAKNIEI DSPYEISRAP DELHYTYLDT FGRPVIVAYK KNLVEQHIQD IVVH
Videos
Documents