missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCT Protein
A cDNA sequence encoding the DCT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11601-10ug
Additional Details : Weight : 0.01000kg
Specifications
1638 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
QFPRVCMTVD SLVNKECCPR LGAESANVCG SQQGRGQCTE VRADTRPWSG PYILRNQDDR ELWPRKFFHR TCKCTGNFAG YNCGDCKFGW TGPNCERKKP PVIRQNIHSL SPQEREQFLG ALDLAKKRVH PDYVITTQHW LGLLGPNGTQ PQFANCSVYD FFVWLHYYSV RDTLLGPGRP YRAIDFSHQG PAFVTWHRYH LLCLERDLQR LIGNESFALP YWNFATGRNE CDVCTDQLFG AARPDDPTLI SRNSRFSSWE TVCDSLDDYN HLVTLCNGTY EGLLRRNQMG RNSMKLPTLK DIRDCLSLQK FDNPPFFQNS TFSFRNALEG FDKADGTLDS QVMSLHNLVH SFLNGTNALP HSAANDPIFV VLHSFTDAIF DEWMKRFNPP ADAWPQELAP IGHNRMYNMV PFFPPVTNEE LFLTSDQLGY SYAIDLPVSV EETPGWPTTH HHHHH | |
Greater than 90% as determined by SDS-PAGE. |
DCT Protein | |
Research Use Only | |
DCT | |
Human | |
His | |
STK3 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |