missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DAG1 Protein

A cDNA sequence encoding the DAG1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11595-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967748

  • £151.37 / 10µg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DAG1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
DAG1 filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASHWPSEPSEAV RDWENQLEAS MHSVLSDLHE AVPTVVGIPD GTAVVGRSFR VTIPTDLIAS SGDIIKVSAA GKEALPSWLH WDSQSHTLEG LPLDTDKGVH YISVSATRLG ANGSHIPQTS SVFSIEVYPE DHSELQSVRT ASPDPGEVVS SACAADEPVT VLTVILDADL TKMTPKQRID LLHRMRSFSE VELHNMKLVP VVNNRLFDMS AFMAGPGNAK KVVENGALLS WKLGCSLNQN SVPDIHGVEA PAREGAMSAQ LGYPVVGWHI ANKKPPLPKR VRR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers