missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cysteine-rich protein 1
A DNA sequence encoding the Homo sapiens (Human) Cysteine-rich protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5868-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001302.1 | |
Cysteine-rich protein 1 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1396 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
CRIP1 | |
Recombinant Protein | |
E. coli | |
PKCPKCNKEV YFAERVTSLG KDWHRPCLKC EKCGKTLTSG GHAEHEGKPY CNHPCYAAMF GPKGFGRGGA ESHTFK |