missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CYP2E1 Protein
A cDNA sequence encoding the CYP2E1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11584-1mg
Additional Details : Weight : 0.01000kg
Specifications
CYP2E1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSWNLPPG PFPLPIIGNL FQLELKNIPK SFTRLAQRFG PVFTLYVGSQ RMVVMHGYKA VKEALLDYKD EFSGRGDLPA FHAHRDRGII FNNGPTWKDI RRFSLTTLRN YGMGKQGNES RIQREAHFLL EALRKTQGQP FDPTFLIGCA PCNVIADILF RKHFDYNDEK FLRLMYLFNE NFHLLSTPWL QLYNNFPSFL HYLPGSHRKV IKNVAEVKEY VSERVKEHHQ SLDPNCPRDL TDCLLVEMEK EKHSAERLYT MDGITVTVAD LFFAGTETTS TTLRYGLLIL MKYPEIEEKL HEEIDRVIGP SRIPAIKDRQ EMPYMDAVVH EIQRFITLVP SNLPHEATRD TIFRGYLIPK GTVVVPTLDS VLYDNQEFPD PEKFKPEHFL NENGKFKYSD YFKPFSTGKR VCAGEGLARM ELFLLLCAIL QHFNLKPLVD PKDIDLSPIH IGFGCIPPRY KLCVIPRS | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CYP2E1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |